siRNA Details
Gene Symbol: Smyd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027188.3 | NP_081464.1 | 2 | 234 | ||
GenBank mRNA | AK010447.1 | - | 2 | 268 | ||
AK043988.1 | - | 2 | 245 | |||
AK087728.1 | - | 2 | 245 | |||
AK163057.1 | - | 2 | 251 | |||
BC019663.1 | - | 2 | 236 | |||
BC038482.1 | - | 2 | 115 | |||
BC052431.1 | - | 2 | 234 | 6837012 | ||
BC061485.1 | - | 2 | 217 | 5370924 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
