siRNA Details
Gene Symbol: Casz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159344.1 | NP_001152816.1 | 5 | 637 | ||
NM_027195.2 | NP_081471.2 | 5 | 637 | |||
GenBank mRNA | BC075673.1 | - | 5 | 639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
