siRNA Details
Gene Symbol: Leng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027203.3 | NP_081479.1 | 1, 2 | 533 | ||
GenBank mRNA | AK005095.1 | - | 1, 2 | 164 | ||
AK037634.1 | - | 1, 2 | 180 | |||
BC023108.1 | - | 1, 2 | 533 | 4021762 |

More Information
Other Species Targeted

Gene Ontology Categories:
