siRNA Details
Gene Symbol: 1500009L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145198.1 | NP_001138670.1 | 6 | 628 | ||
GenBank mRNA | AK005183.1 | - | 6 | 559 | ||
AK012407.1 | - | 590 | ||||
BC048550.1 | - | 6 | 628 |
