siRNA Details
Gene Symbol: Ceacam13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027210.3 | NP_081486.1 | 2 | 187 | ||
NM_028171.2 | NP_082447.1 | 2 | 187 | |||
GenBank mRNA | AK005430.1 | - | 2 | 178 | ||
AK005537.1 | - | 2 | 188 | |||
AK146045.1 | - | 2 | 188 | |||
BC099430.1 | - | 2 | 174 | 30787167 | ||
BY705748.1 | - | 2 | 188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
