siRNA Details
Gene Symbol: Med30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027212.2 | NP_081488.1 | 4 | 544 | ||
GenBank mRNA | AK007739.1 | - | 4 | 537 | ||
AK011104.1 | - | 4 | 545 | |||
AK146250.1 | - | 4 | 549 | |||
BC020113.1 | - | 4 | 532 | 4015173 |

More Information
Other Species Targeted

Gene Ontology Categories:
