siRNA Details
Gene Symbol: Cox11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199008.2 | NP_950173.1 | 3 | 636 | ||
GenBank mRNA | AK008096.1 | - | 3 | 650 | ||
AK143937.1 | - | 3 | 636 | |||
AK172067.1 | - | 3 | 640 | |||
BC061233.1 | - | 3 | 647 | 6585499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
