siRNA Details
Gene Symbol: 1810046K07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027217.1 | NP_081493.1 | 4 | 331 | ||
GenBank mRNA | AK007796.1 | - | 4 | 334 | ||
BC120857.1 | - | 2 | 115 | 40129780 | ||
BC120859.1 | - | 2 | 115 | 40129782 |
