siRNA Details
Gene Symbol: Clec4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190310.1 | NP_001177239.1 | 6 | 711 | ||
NM_027218.3 | NP_081494.1 | 5 | 612 | |||
GenBank mRNA | AK007794.1 | - | 5 | 569 | ||
AY230259.1 | - | 6 | 675 | |||
AY230260.1 | - | 5 | 576 | |||
AY365134.1 | - | 6 | 607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
