siRNA Details
Gene Symbol: Mzb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027222.3 | NP_081498.2 | 2 | 238 | ||
GenBank mRNA | AK008016.1 | - | 2 | 239 | ||
AK088094.1 | - | 2 | 258 | |||
BC030674.1 | - | 1 | 20 | |||
BC064044.1 | - | 2 | 225 | 875322 | ||
GQ477351.1 | - | 2 | 204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
