siRNA Details
Gene Symbol: Mterf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178051.3 | NP_835152.1 | 3 | 714 | ||
GenBank mRNA | AK007901.1 | - | 3 | 701 | ||
AK077142.1 | - | 3 | 714 | |||
AK079098.1 | - | 3 | 706 | |||
BC002280.1 | - | 2 | 327 | 3497421 | ||
BC055877.1 | - | 2 | 658 | 4236729 | ||
BC064709.1 | - | 3 | 707 | 30314353 |
