siRNA Details
Gene Symbol: Ms4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023529.2 | NP_076018.2 | 5, 6 | 560 | ||
GenBank mRNA | AB026046.1 | - | 5, 6 | 569 | ||
AF237918.1 | - | 5, 6 | 560 | |||
AK008019.1 | - | 3, 4 | 419 | |||
BC144935.1 | - | 5, 6 | 557 | 9053483 | ||
BC150675.1 | - | 5, 6 | 557 | 9087586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
