siRNA Details
Gene Symbol: Polr2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027231.1 | NP_081507.1 | 3 | 217 | ||
GenBank mRNA | AK007913.1 | - | 3 | 232 | ||
AK165600.1 | - | 3 | 244 | |||
AK169434.1 | - | 3 | 233 | |||
BC024419.1 | - | 3 | 217 | 4983359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
