siRNA Details
Gene Symbol: Pcgf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197992.1 | NP_932109.1 | 7 | 633 | ||
GenBank mRNA | AK008048.1 | - | 7 | 615 | ||
BC028560.1 | - | 7 | 633 | 1380201 |

More Information
Other Species Targeted

Gene Ontology Categories:
