siRNA Details
Gene Symbol: 2010003K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027237.1 | NP_081513.1 | 2 | 642 | ||
GenBank mRNA | AK008072.1 | - | 2 | 620 | ||
BC021951.1 | - | 2 | 642 | 5102159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
