siRNA Details
Gene Symbol: Mfsd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178620.3 | NP_848735.1 | 13 | 1348 | ||
GenBank mRNA | AK031384.1 | - | 13 | 1543 | ||
AK032043.1 | - | 13 | 1544 | |||
AK191123.1 | - | 2 | 120 | |||
BC016209.1 | - | 14 | 1307 | |||
BC079583.1 | - | 13 | 1375 | |||
BC088988.1 | - | 13 | 1348 | 6418757 | ||
BC100547.1 | - | 13 | 3424 | |||
BC137970.1 | - | 13 | 1335 | 8860989 | ||
BC137971.1 | - | 13 | 1336 | 8860990 | ||
BC145068.1 | - | 13 | 1315 |
