siRNA Details
Gene Symbol: Rasip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028544.1 | NP_082820.1 | 8 | 2164 | ||
GenBank mRNA | AK011557.2 | - | 2 | 247 | ||
AK033276.1 | - | 6 | 1452 | |||
AK156602.1 | - | 8 | 2176 | |||
BC028444.1 | - | 236 | ||||
BC072584.1 | - | 5 | 1118 | |||
BC127059.1 | - | 6 | 1090 | 40109852 | ||
BC150766.1 | - | 8 | 2164 | 9087677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
