siRNA Details
Gene Symbol: Nup43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145706.2 | NP_663752.2 | 8 | 1077 | ||
GenBank mRNA | AK011422.1 | - | 8 | 1041 | ||
AK012191.1 | - | 8 | 1071 | |||
AK090019.1 | - | 8 | 1040 | |||
AK146262.1 | - | 8 | 1079 | |||
BC011060.1 | - | 8 | 1065 | 3156436 | ||
BC052530.1 | - | 8 | 1005 | |||
DV046453.1 | - | 550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
