siRNA Details
Gene Symbol: Polr2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027259.1 | NP_081535.1 | 3 | 429 | ||
GenBank mRNA | AB221537.1 | - | 3 | 157 | ||
AK012635.1 | - | 3 | 166 | |||
BC062812.1 | - | 3 | 429 | 3466417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
