siRNA Details
Gene Symbol: Agk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023538.2 | NP_076027.1 | 4 | 259 | ||
GenBank mRNA | AJ401619.1 | - | 4 | 232 | ||
AK011715.1 | - | 4 | 255 | |||
AK051531.1 | - | 1 | 2411 | |||
AK076224.1 | - | 3 | 222 | |||
AK155346.1 | - | 4 | 225 | |||
AY772469.1 | - | 3 | 200 | |||
BC019145.1 | - | 4 | 253 | 5051535 | ||
BC093525.1 | - | 4 | 229 | 5151866 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
