siRNA Details
Gene Symbol: Fars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039189.2 | NP_001034278.1 | 5 | 943 | ||
NM_024274.3 | NP_077236.1 | 6 | 1576 | |||
GenBank mRNA | AK013211.1 | - | 6 | 1315 | ||
AK020164.1 | - | 5 | 729 | |||
AK054370.1 | - | 1342 | ||||
AK076259.1 | - | 6 | 1361 | |||
AK170986.1 | - | 6 | 1359 | |||
BC002147.1 | - | 6 | 1346 | 3484842 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
