siRNA Details
Gene Symbol: Spink2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289764.1 | NP_001276693.1 | 2 | 106 | ||
NM_001289767.1 | NP_001276696.1 | 2 | 202 | |||
NM_001289768.1 | NP_001276697.1 | 2 | 185 | |||
NM_183284.3 | NP_899107.1 | 2 | 185 | |||
GenBank mRNA | AK005709.1 | - | 2 | 168 | ||
BC048619.1 | - | 2 | 152 | |||
BC087876.1 | - | 2 | 148 | 6773100 | ||
DV068802.1 | - | 2 | 205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
