siRNA Details
Gene Symbol: 1700028J19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006462.1 | - | 3 | 359 | ||
BC049653.1 | - | 3 | 324 | |||
BC061239.1 | - | 2 | 239 | |||
BC087938.1 | - | 2 | 239 | |||
BC100477.1 | - | 3 | 340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
