siRNA Details
Gene Symbol: Ace2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130513.1 | NP_001123985.1 | 6 | 1017 | ||
NM_027286.4 | NP_081562.2 | 5 | 869 | |||
GenBank mRNA | AB053181.1 | - | 5 | 873 | ||
AB053182.1 | - | 5 | 873 | |||
AK141604.1 | - | 5 | 718 | |||
BC026801.1 | - | 5 | 862 | 4236529 | ||
EF408739.1 | - | 6 | 978 | |||
EF408740.1 | - | 6 | 978 | |||
GQ262785.1 | - | 5 | 687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
