siRNA Details
Gene Symbol: Ino80b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023547.1 | NP_076036.1 | 3 | 298 | ||
GenBank mRNA | AK010946.1 | - | 3 | 400 | ||
BC019383.1 | - | 3 | 332 | 5010975 | ||
BC047990.1 | - | 3 | 333 | |||
BC058788.1 | - | 3 | 344 | 6440887 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
