siRNA Details
Gene Symbol: Acot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146057.1 | NP_001139529.1 | 4 | 539 | ||
NM_001146058.1 | NP_001139530.1 | 4 | 401 | |||
NM_133348.2 | NP_579926.2 | 4 | 528 | |||
GenBank mRNA | AB049821.1 | - | 4 | 494 | ||
AB088411.1 | - | 4 | 467 | |||
AB088412.1 | - | 4 | 435 | |||
AB207243.1 | - | 4 | 494 | |||
AK152277.1 | - | 4 | 409 | |||
AK198843.1 | - | 2 | 38 | |||
AK213487.1 | - | 2 | 38 | |||
BC013507.1 | - | 4 | 396 | 4224774 | ||
CA463703.1 | - | 531 | ||||
CX235974.1 | - | 4 | 420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
