siRNA Details
Gene Symbol: Cmtm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027294.2 | NP_081570.1 | 4 | 702 | ||
GenBank mRNA | AK012262.1 | - | 4 | 703 | ||
AY243513.1 | - | 4 | 737 | |||
BC027554.1 | - | 4 | 624 | 1382818 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
