siRNA Details
Gene Symbol: Ifi35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027320.4 | NP_081596.1 | 3 | 397 | ||
GenBank mRNA | AK008161.1 | - | 3 | 329 | ||
AK131884.1 | - | 3 | 398 | |||
BC008158.1 | - | 3 | 301 | 3155505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
