siRNA Details
Gene Symbol: Odf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013022.1 | NP_001013040.1 | 5 | 644 | ||
GenBank mRNA | AK008010.1 | - | 4 | 536 | ||
BC087962.1 | - | 5 | 644 | 6748887 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
