siRNA Details
Gene Symbol: Mllt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027326.3 | NP_081602.3 | 10, 11 | 1895 | ||
NM_029931.3 | NP_084207.2 | 6, 7 | 568 | |||
GenBank mRNA | AF333960.1 | - | 9, 10 | 1556 | ||
AK008707.1 | - | 6, 7 | 530 | |||
AK136066.1 | - | 6, 7 | 1129 | |||
AK147009.1 | - | 6, 7 | 482 | |||
BC021420.1 | - | 11, 12 | 1734 | 5012671 | ||
BU614509.1 | - | 6, 7 | 696 | |||
DV051568.1 | - | 578 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053718, AJ006295, BC129089.
Gene Ontology Categories:
