siRNA Details
Gene Symbol: Mllt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286158.1 | NP_001273087.1 | 6 | 525 | ||
NM_027326.3 | NP_081602.3 | 11 | 1924 | |||
NM_029931.3 | NP_084207.2 | 7 | 597 | |||
GenBank mRNA | AF333960.1 | - | 10 | 1585 | ||
AK008707.1 | - | 7 | 559 | |||
AK019458.1 | - | 6 | 517 | |||
AK136066.1 | - | 7 | 1158 | |||
AK147009.1 | - | 7 | 511 | |||
BC021420.1 | - | 12 | 1763 | 5012671 | ||
BU614509.1 | - | 7 | 725 | |||
BY713607.1 | - | 6 | 513 | |||
DV051568.1 | - | 607 |
