siRNA Details
Gene Symbol: 2210013O21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027327.1 | NP_081603.1 | 3 | 584 | ||
GenBank mRNA | AK008724.1 | - | 3 | 550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
