siRNA Details
Gene Symbol: Slc44a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023557.3 | NP_076046.1 | 8 | 575 | ||
GenBank mRNA | AK145621.1 | - | 8 | 567 | ||
AK164684.1 | - | 7 | 1343 | |||
AK166620.1 | - | 5 | 477 | |||
BC010808.1 | - | 8 | 573 | 4240658 | ||
BC052652.1 | - | 8 | 573 | 30034507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
