siRNA Details
Gene Symbol: Mettl7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027334.3 | NP_081610.2 | 2 | 906 | ||
GenBank mRNA | AJ536232.1 | - | 2 | 878 | ||
AK076169.1 | - | 2 | 908 | |||
BC033446.1 | - | 2 | 883 | |||
BC047149.1 | - | 2 | 891 | |||
BC055034.1 | - | 2 | 897 | 6306873 | ||
BC060144.1 | - | 2 | 886 |
