siRNA Details
Gene Symbol: Vps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027338.1 | NP_081614.1 | 8 | 603 | ||
GenBank mRNA | AK150754.1 | - | 8 | 624 | ||
AK151035.1 | - | 8 | 624 | |||
AK156241.1 | - | 617 | ||||
AK161865.1 | - | 655 | ||||
BC010811.1 | - | 8 | 603 | 4210911 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
