siRNA Details
Gene Symbol: Lipn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027340.2 | NP_081616.1 | 7 | 959 | ||
GenBank mRNA | AK008974.1 | - | 6 | 589 | ||
AK154333.1 | - | 7 | 960 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
