siRNA Details
Gene Symbol: Abhd17c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133722.2 | NP_598483.2 | 2 | 761 | ||
GenBank mRNA | BC018511.1 | - | 2 | 761 | 3483379 | |
BM943880.1 | - | 2 | 242 |

More Information
Other Species Targeted

Gene Ontology Categories:
