siRNA Details
Gene Symbol: Gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027352.4 | NP_081628.3 | 4, 5 | 620 | ||
GenBank mRNA | AK002515.1 | - | 5, 6 | 699 | ||
AK010664.2 | - | 4, 5 | 539 | |||
AK033413.1 | - | 4, 5 | 554 | |||
AK044691.1 | - | 505 | ||||
AK077153.1 | - | 4, 5 | 495 | |||
AK077683.1 | - | 4, 5 | 555 | |||
AK077706.1 | - | 4, 5 | 487 | |||
AK088239.1 | - | 4, 5 | 501 | |||
AK148337.1 | - | 4, 5 | 554 | |||
AK151843.1 | - | 4, 5 | 502 | |||
AK152475.1 | - | 4, 5 | 502 | |||
AK152544.1 | - | 4, 5 | 502 | |||
AK152603.1 | - | 4, 5 | 530 | |||
AK153049.1 | - | 4, 5 | 502 | |||
AK153555.1 | - | 4, 5 | 553 | |||
AK159469.1 | - | 4, 5 | 552 | |||
AK167823.1 | - | 4, 5 | 554 | |||
AK168928.1 | - | 5, 6 | 750 | |||
AK169382.1 | - | 4, 5 | 502 | |||
AK207507.1 | - | 2, 3 | 99 | |||
AK212581.1 | - | 2, 3 | 99 | |||
AK216052.1 | - | 2, 3 | 159 | |||
BC005600.1 | - | 4, 5 | 493 | 3482776 | ||
BC016455.1 | - | 3, 4 | 345 | 4234343 | ||
BC024982.1 | - | 4, 5 | 506 | |||
BU707592.1 | - | 4, 5 | 620 |

More Information
Other Species Targeted

Gene Ontology Categories:
