siRNA Details
Gene Symbol: Rnf168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027355.2 | NP_081631.2 | 1 | 759 | ||
GenBank mRNA | AK013946.1 | - | 1 | 719 | ||
AK053769.1 | - | 1 | 762 | |||
BC046815.1 | - | 1 | 731 | 5702430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
