siRNA Details
Gene Symbol: Ufsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027356.2 | NP_081632.1 | 1 | 234 | ||
GenBank mRNA | AK012343.1 | - | 1 | 235 | ||
BC016577.1 | - | 1 | 243 | 3584484 | ||
BC087958.1 | - | 1 | 255 | 6751787 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
