siRNA Details
Gene Symbol: 2010107E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027360.2 | NP_081636.1 | 2 | 154 | ||
GenBank mRNA | AK008333.1 | - | 2 | 155 | ||
AK020400.1 | - | 1 | 570 | |||
BC055712.1 | - | 2 | 133 | 6408438 | ||
BC059719.1 | - | 2 | 142 | 6774233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
