siRNA Details
Gene Symbol: Rnf126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144528.3 | NP_653111.1 | 4 | 388 | ||
GenBank mRNA | AK042319.1 | - | 4 | 255 | ||
AK154781.1 | - | 4 | 354 | |||
BB870939.1 | - | 4 | 357 | |||
BC016543.1 | - | 4 | 360 | 3158083 | ||
BU147682.1 | - | 4 | 452 | |||
DV648665.1 | - | 4 | 283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
