siRNA Details
Gene Symbol: Rnf126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144528.3 | NP_653111.1 | 9 | 990 | ||
GenBank mRNA | AK042319.1 | - | 8 | 787 | ||
AK154781.1 | - | 9 | 956 | |||
BC016543.1 | - | 9 | 962 | 3158083 | ||
CA321804.1 | - | 6 | 497 |
