siRNA Details
Gene Symbol: Iyd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027391.3 | NP_081667.1 | 3 | 456 | ||
GenBank mRNA | AK002363.1 | - | 3 | 457 | ||
BC023358.1 | - | 3 | 479 | 5064638 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
