siRNA Details
Gene Symbol: Fbln7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024237.4 | NP_077199.2 | 7, 8 | 1095 | ||
GenBank mRNA | AK005465.1 | - | 7, 8 | 1047 | ||
BC002308.1 | - | 2, 3 | 171 | |||
BC051550.1 | - | 5, 6 | 651 | |||
BC095941.1 | - | 7, 8 | 1027 | 4167912 | ||
BM936626.1 | - | 3, 4 | 356 | |||
EF668007.1 | - | 7, 8 | 951 |
