siRNA Details
Gene Symbol: Fbln7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024237.4 | NP_077199.2 | 2 | 344 | ||
GenBank mRNA | AK005465.1 | - | 2 | 299 | ||
BB867118.1 | - | 2 | 358 | |||
BC095941.1 | - | 2 | 276 | 4167912 | ||
CK023294.1 | - | 2 | 286 | |||
EF668007.1 | - | 2 | 203 |
