siRNA Details
Gene Symbol: Gpatch2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027405.2 | NP_081681.2 | 3 | 793 | ||
GenBank mRNA | AK006185.1 | - | 3 | 794 | ||
AK031712.1 | - | 3 | 797 | |||
AK129299.1 | - | 3 | 770 | |||
AK147916.1 | - | 3 | 782 | |||
BC044824.1 | - | 2 | 113 | |||
BC048169.1 | - | 3 | 802 | 6821902 | ||
BC050782.1 | - | 3 | 773 | |||
BC058256.1 | - | 3 | 793 | 6466603 |
