siRNA Details
Gene Symbol: Gpatch2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027405.2 | NP_081681.2 | 9 | 1296 | ||
GenBank mRNA | AK129299.1 | - | 9 | 1273 | ||
AK147916.1 | - | 9 | 1285 | |||
BC048169.1 | - | 9 | 1305 | 6821902 | ||
BC050782.1 | - | 9 | 1276 | |||
BC058256.1 | - | 9 | 1296 | 6466603 |
