siRNA Details
Gene Symbol: Tecpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027410.1 | NP_081686.1 | 22 | 3253 | ||
GenBank mRNA | AB093295.1 | - | 21 | 3422 | ||
AK008701.1 | - | 5 | 491 | |||
AK032858.1 | - | 22 | 3227 | |||
AK169888.1 | - | 22 | 3200 | |||
AK170108.1 | - | 22 | 3200 | |||
AK195442.1 | - | 2 | 111 | |||
AK195977.1 | - | 2 | 111 | |||
AK202952.1 | - | 2 | 194 | |||
AK211310.1 | - | 2 | 220 | |||
AK219493.1 | - | 2 | 220 | |||
BC004058.1 | - | 5 | 539 | |||
BC025567.1 | - | 9 | 998 | |||
BC046968.1 | - | 22 | 3253 | 6306206 |
