siRNA Details
Gene Symbol: Asnsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290984.1 | NP_001277913.1 | 5 | 578 | ||
NM_133728.3 | NP_598489.2 | 5 | 2009 | |||
GenBank mRNA | AK031295.1 | - | 5 | 1967 | ||
AK034746.1 | - | 1928 | ||||
AK049194.1 | - | 5 | 1956 | |||
AK050045.1 | - | 5 | 1949 | |||
AK076793.1 | - | 5 | 1911 | |||
AK155030.1 | - | 5 | 1948 | |||
AK168103.1 | - | 5 | 1949 | |||
AK168534.1 | - | 5 | 579 | |||
AK170510.1 | - | 5 | 1925 | |||
AK176998.1 | - | 1 | 52 | |||
AK205803.1 | - | 1 | 33 | |||
AK218145.1 | - | 1 | 34 | |||
BC013617.1 | - | 5 | 1829 | 3979452 |

More Information
Other Species Targeted

Gene Ontology Categories:
